Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Supplier:   Ludwig Schneider
Density hydrometers
Supplier:  Ludwig Schneider
Certificates
Density hydrometers
12 Options Available Below
Product Details & Documents
Precision hydrometers with integrated blue filling thermometer.
  • Suitable for calibration
Error limit: ±0,5 kg/m³
Reference temperature: 15 °C
Certifications: ASTM E2995-14
Ordering information: A works or DAkkS calibration certificate is available on request.
Packaging: Each packed in a square plastic sleeve.
Specifications for Product Family
Specifications
  • Catalog No:
  • SCNL3222111S
  • SCNL3222112S
  • SCNL3222113S
  • SCNL3222114S/04/3P
  • SCNL3222114S
  • SCNL3222115S
  • SCNL3222116S/04/3P
  • SCNL3222116S
  • SCNL3222117S
  • SCNL3222118S
  • SCNL3222119S
  • SCNL3222120S
  • Reference temperature:
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
  • 15 °C
Specifications
Product Family Options
Product Information
  • Model
    Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • S500 HL-14
    0,600 - 0,650 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222111S
    Reference temperature
    15 °C
  • S501 HL-14
    0,650 - 0,700 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222112S
    Reference temperature
    15 °C
  • S502 HL-14
    0,700 - 0,750 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222113S
    Reference temperature
    15 °C
  • S503 HL-14
    0,750 - 0,800 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 606,00 €
    606,00 €
    1 * 1 ST
       606,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222114S/04/3P
    Reference temperature
    15 °C
  • S503 HL-14
    0,750 - 0,800 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222114S
    Reference temperature
    15 °C
  • S504 HL-14
    0,800 - 0,850 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222115S
    Reference temperature
    15 °C
  • S505 HL-14
    0,850 - 0,900 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 606,00 €
    606,00 €
    1 * 1 ST
       606,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222116S/04/3P
    Reference temperature
    15 °C
  • S505 HL-14
    0,850 - 0,900 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222116S
    Reference temperature
    15 °C
  • S506 HL-14
    0,900 - 0,950 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222117S
    Reference temperature
    15 °C
  • S507 HL-14
    0,950 - 1,000 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,00 €
    83,00 €
    1 * 1 ST
       83,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222118S
    Reference temperature
    15 °C
  • S508 HL-14
    1,000 - 1,050 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222119S
    Reference temperature
    15 °C
  • S509 HL-14
    1,050 - 1,100 g/ml
    0,0005 g/ml
    −20...+65 °C in 1 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3222120S
    Reference temperature
    15 °C