Order Entry
Belgium
ContactUsLinkComponent
 
Bovine serum albumin (BSA)
 :  SEQENS
 
undefined
Bovine serum albumin (BSA)
 :  SEQENS

 

  • Source:
    Serum
  • Species:
    Bovine
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Endotoxin level:
    very low
  • Concentration:
    30% solution

 

Frequently Bought Together