Order Entry
Belgium
ContactUsLinkComponent
Genomic RNA purification kits, Fast RNA™ Pro Red kit, MP Biomedicals
Genomic RNA purification kits, Fast RNA™ Pro Red kit, MP Biomedicals
Catalog # ICNA116035050
Supplier:  MP Biomedicals
CAS Number:  
undefined
Genomic RNA purification kits, Fast RNA™ Pro Red kit, MP Biomedicals
Catalog # ICNA116035050
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Yeast/Fungal RNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    RNA
  • Description:
    Fast RNA™ Pro Red Kit, 50×2 ml preps
  • Description:
    Fast RNA™ Pro Red Kit

Specifications

About this item

FastRNA® Pro Red Kit is used to isolate total RNA from yeast and fungi. Isolated RNA is suitable for RT-PCR analysis and other downstream applications

Using the FastPrep-24™ or FastPrep® FP120 instrument, up to 10⁹ cells are homogenised by Lysing Matrix C in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.

For research purposes only.

Delivery: Kit contains RNApro™ Solution (1×55 ml), DEPC-treated H₂O (1×15 ml) and Lysing Matrix C (50×2 ml tubes).