Bovine serum albumin (BSA)
: SEQENS
SEQENS IVD manufactures Bovine Serum Albumin (BSA) derived from plasma, collected from healthy animals of French origin. This product is obtained by 'Heat Shock' method, which ensures higher purity and preserves BSA intrinsic properties.
- Albumin purity: >97%
- Total proteins: >290 g/L
- Proteases: Undetectable
- Shelf life: 3 years
- Sterile filtered 0,2 µm
Full traceability and batch-to-batch repeatability guaranteed.
: Supplied as a ready to use 30% solution in deionised water.
- Source:Serum
- Species:Bovine
- Protease-free:Y
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Concentration:30% solution
Frequently Bought Together