ContactUsLinkComponent
Bulk adsorbents, Florisil® PR
Bulk adsorbents, Florisil® PR
Catalog # REST26135
Supplier:  Restek
CAS Number:  
undefined
Bulk adsorbents, Florisil® PR
Catalog # REST26135
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Florisil® PR bulk adsorbent, 60/100 mesh
Specifications
About this item
Florisil® PR is commonly used to remove polar interferences from pesticide residues. This bulk material is ideal for labs packing their own extraction cartridges for pesticide residue extractions.
  • Pesticide residue grade
Packaging: Packaged in glass containers.