Order Entry
Belgium
ContactUsLinkComponent
 
Residual chlorine standard for ISE calibration, Orion™
Catalog # 662-0880
Supplier:  Thermo Orion
CAS Number:  
Residual chlorine standard for ISE calibration, Orion™
Catalog # 662-0880
Supplier:  Thermo Orion
CAS Number:  

Specifications

  • Concentration:
    100 ppm
  • Pk:
    475 mL
  • Description:
    Residual chlorine standard for ISE calibration

Specifications

About this item