Order Entry
Belgium
ContactUsLinkComponent
Water hardness standard for ISE calibration, Orion™
Water hardness standard for ISE calibration, Orion™
Catalog # 662-0806
Supplier:  Thermo Orion
CAS Number:  
Water hardness standard for ISE calibration, Orion™
Catalog # 662-0806
Supplier:  Thermo Orion
CAS Number:  

Specifications

  • Concentration:
    100 ppm CaCO₃
  • Pk:
    475 mL
  • Description:
    Water hardness standard for ISE calibration

Specifications

About this item