Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Bromide Standards
Bromide standard for ISE calibration, Orion™
Bromide standard for ISE calibration, Orion™
Bromide standard for ISE calibration, Orion™
Catalog # ORIO943506
Supplier:
Thermo Orion
CAS Number:
Expand image
Bromide standard for ISE calibration, Orion™
Catalog # ORIO943506
Supplier:
Thermo Orion
CAS Number:
Specifications
Concentration:
0,1M
Pk:
475 mL
Description:
Bromide standard solution
Specifications
About this item
Product Details
Product Details