Order Entry
Belgium
ContactUsLinkComponent
 
Ion selective electrodes, cadmium, Orion™
Catalog # 662-1572
Supplier:  Thermo Orion
undefined
Ion selective electrodes, cadmium, Orion™
Catalog # 662-1572
Supplier:  Thermo Orion

Specifications

  • Concentration:
    0,1 M - 10-7 M
  • Connection:
    Waterproof BNC, 1 m cable
  • Type:
    9648BNWP
  • Description:
    Cadmium solid-state combination electrode, ionplus® 9648BNWP
  • Seq:
    03
  • Pk:
    1
  • Temp.:
    0...+80 °C

Specifications

About this item

These electrodes are designed for measuring cadmium ions in aqueous solutions quickly, simply and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode (9648BNWP) does not require a separate reference electrode, making it convenient to use with small sample volumes
Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.

Reference electrode

Product Information