Orders LinkContactUsLinkComponent
pH software, EasyDirect™
License key for EasyDirect™ pH software
  661-0271
pH software, EasyDirect™
  661-0271
pH software, EasyDirect™
pH software, EasyDirect™
License key for EasyDirect™ pH software
265,00 €
1 * 1 ST
This intuitive software for improved data management can be used with SevenCompact™, SevenCompact™ Duo, Seven2Go™ Pro, FiveEasyPlus or SevenExcellence™ pH meters. Within a few clicks the data measured on the pH meter can be seamlessly transferred to a computer. The application will store the data unmodified and allow for further printing or analysis.
  • Available in 12 languages
  • Data can be stored, printed or exported to Excel or any other csv application
  • Up to three instruments can be connected simultaneously
 : Supplied without meter, computer and USB cable.
  • Description:
    License key for EasyDirect™ pH software
  • Seq:
    40
Accessories
  •  
    Description
  • 661-0103
    USB cable 1,8 m for PC or printer
    1 * 1 ST - 30,20 €