Order Entry
Belgium
ContactUsLinkComponent
Dow Corning® SILASTIC S
Dow Corning® SILASTIC S
Catalog # 634712A
Supplier:  DOW CORNING
CAS Number:  
Dow Corning® SILASTIC S
Catalog # 634712A
Supplier:  DOW CORNING
CAS Number:  

Specifications

  • Pk:
    1,1 kg
  • Seq:
    4038
  • Environmentally Preferable:

Specifications

About this item

 :