Anti-Msx2 Mouse Monoclonal Antibody [clone: 2E12]
Catalog # ENZOADIAAM010E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Msh Homeobox 2
- Antigen symbol:MSX2
- Clonality:Monoclonal
- Clone:2E12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Epitope:N-Terminal
- Form:Liquid
- Antigen synonyms:MSH|FPP|HOX8|PFM|PFM1|CRS2
- Storage buffer:PBS, pH 7,2, containing 50% glycerol.
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Msx2 (N-terminal fragment)
- Purification:Purified by Protein A affinity chromatography
- Pk:100 µG
Specifications
About this item
Msx2, also known as HOX-8, is a homeobox containing transcription factor, which is a member of the MSH family of homeobox proteins.
- Dilution Range: 10 µg/ml for immunohistochemistry, 1 µg/ml for Western blot
Homeobox (HOX) genes encode DNA binding proteins that recognize specific sequences and modulate transcriptional activity. Msx2 is believed to play a role in regulating bone development, particularly in sutures of cranial bone.
Type: Primary
Antigen: Msh Homeobox 2
Clonality: Monoclonal
Clone: 2E12
Conjugation:
Epitope: N-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human