Order Entry
Belgium
Orders LinkContactUsLinkComponent
α-Cyclodextrin, white powder

α-Cyclodextrin, white powder

Supplier:   MP Biomedicals
α-Cyclodextrin, white powder
Supplier:  MP Biomedicals
Certificates

2 Options Available Below

Product Details & Documents

α-Cyclodextrin is reported useful for the selective precipitation of enantiomeric, positional or structural isomers. Cyclodextrins have hydrophobic cavities; form inclusion compounds with organic substances, salts and halogens in the solid state or in aqueous solutions.

Soluble in water (50 mg/ml).

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Melting Pt:278 °C (lit.)
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Identity Test:
    Passes
  • Loss on Drying:
    ≤13%
  • Optical Rotation:
    +148 ± 5°(c=1, water)
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 80,00 €
    80,00 €
    1 * 5 g
       80,00 €

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 361,00 €
    361,00 €
    1 * 25 g
       361,00 €