Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Phenylethanol, liquid

2-Phenylethanol, liquid

1 L
Catalog # ICNA0215617691
Corrosive
Toxic
Supplier:   MP Biomedicals
2-Phenylethanol, liquid
Catalog # ICNA0215617691
Supplier:  MP Biomedicals
CAS Number:  60-12-8
2-Phenylethanol, liquid

1 L

CAS Number:  60-12-8
Price
97,00 €
Log in for availability
1 * 1 L
Product Details & Documents

2-Phenylethanol, an alcohol potentially used as an anti-microbial.

Soluble in most fixed oils, common organic solvents, water and propylene glycol.

Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Melting Pt:–27 °C
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.