Order Entry
Belgium
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%
Supplier:  MP Biomedicals
Certificates
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥95%
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    β-NADPH; TPNH; Coenzyme II reduced form tetrasodium salt; Dihydronicotinamide-adenine dinucleotide phosphate tetrasodium salt; Triphosphopyridine nucleotide reduced tetrasodium salt
  • Form:
    White Powder
  • Storage:
    Store at -0 °C, Desiccate, Protect from light, Store under nitrogen

Specification Test Results

Frequently Bought Together