Carboxymethyl cellulose sodium salt, 200 - 800 cps, powder, medium viscosity
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9004-32-4
:
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Viscosity (2% aq soln):200 - 800 cps
Specification Test Results
Frequently Bought Together