Hypoxanthine disodium salt ≥98.0% (dried basis), off-white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:68-94-0
Formula:C₅H₂N₄Na₂O
MDL Number:MFCD00005725
Melting Pt:>300 °C (lit.)
:180.08 g/mol
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Reference
- UV/Vis: l max (0.1 N HCl ):246 - 249 nm
- OD250/OD260:1.40 - 1.55
- OD280/OD260:0.01 - 0.15
- pH (1% aq soln):11.5 - 12.5
- Assay (on dried basis) :≥98.0%
Specification Test Results
Frequently Bought Together