Order Entry
Belgium
ContactUsLinkComponent
 
L(+)-Rhamnose monohydrate ∼99%, powder
 
L(+)-Rhamnose monohydrate ∼99%, powder
 

 

  • Identity Test :
    Passes
  • Purity:
    ~99%
  • FTIR:
    Conforms to Standard
  • Melting Point:
    91 - 101 °C
  • Moisture:
    Report Result
  • Optical Rotation (c=4, H₂O, NH₃):
    +8.2 ± 1°

 

 

Frequently Bought Together