Order Entry
Belgium
ContactUsLinkComponent
Sodium deoxycholate, white powder
Irritant
Sodium deoxycholate, white powder
Supplier:  MP Biomedicals
Certificates
Irritant
Sodium deoxycholate, white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • pH (2% aq soln):
    7.5-9.5
  • Loss on Drying:
    <5%
  • Sodium Cholate:
    ≤2%
  • Purity:
    Report Result

Specification Test Results

Frequently Bought Together