Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(+)-Glutamine

L(+)-Glutamine

Glass bottle, 100 mL
Catalog # ICNA091680149
Supplier:   MP Biomedicals
L(+)-Glutamine
Catalog # ICNA091680149
Supplier:  MP Biomedicals
CAS Number:  56-85-9
L(+)-Glutamine
L(+)-Glutamine

Glass bottle, 100 mL

CAS Number:  56-85-9
Price
28,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications