Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98,5%, white powder
Irritant
Health
tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98,5%, white powder
Supplier:  MP Biomedicals
Certificates
Irritant
Health
tert-Butyl-4-methoxyphenol (mixture of isomers) ∼98,5%, white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ~98.5%
  • FTIR:
    Conforms to Reference

Specification Test Results

Frequently Bought Together