Mouse Prolactin ELISA kit
Catalog # BIRBORB138058-96
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:96
- Primary antibody reactivity:Mouse
- Description:Mouse Prolactin ELISA Kit
- Detection range:156 - 10000 pg/ml
- Sensitivity:<10 pg/ml
Specifications
About this item
Mouse Prolactin ELISA Kit for the quantitative detection of mouse prolactin in cell culture supernates, serum and plasma (heparin, citrate).