Order Entry
Belgium
ContactUsLinkComponent
Human TIMP-4 ELISA kit
Human TIMP-4 ELISA kit
Catalog # BIRBORB138055-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human TIMP-4 ELISA kit
Catalog # BIRBORB138055-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    TIMP-4
  • Description:
    Human TIMP-4 ELISA Kit
  • Detection range:
    78 - 5000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human TIMP-4 ELISA Kit for the quantitative detection of human Tissue inhibitor of metalloproteinases 4 (TIMP-4) in cell culture supernates, serum, plasma (heparin, EDTA) and human milk.

Tissue inhibitor of metalloproteinases 4, also called Metalloproteinase inhibitor 4, is an enzyme that in humans is encoded by the TIMP4 gene. This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. This gene is mapped to 3p25.2. The secreted, netrin domain-containing protein encoded by this gene is involved in regulation of platelet aggregation and recruitment and may play role in hormonal regulation and endometrial tissue remodeling.