ContactUsLinkComponent
Mouse E-selectin ELISA kit
  BIRBORB99236-96
 :  Biorbyt
 :  
undefined
Mouse E-selectin ELISA kit
  BIRBORB99236-96
 :  Biorbyt
 :  
  • Pk:
    96
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse E-Selectin ELISA Kit
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml
Mouse E-Selectin ELISA Kit for the quantitative detection of mouse soluble E-Selectin in cell culture supernates, serum and plasma (heparin, citrate).
  • No detectable cross-reactivity with other relevant proteins