Order Entry
Belgium
ContactUsLinkComponent
 
Human IL-22 ELISA kit
  BIRBORB99233-96
 :  Biorbyt
 :  
undefined
Human IL-22 ELISA kit
  BIRBORB99233-96
 :  Biorbyt
 :  

 

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-22
  • Description:
    Human IL-22 ELISA Kit
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <10 pg/ml

 

 

Human IL-22 ELISA Kit for the quantitative detection of human Interleukin-22 in cell culture supernates, serum and plasma (heparin, EDTA).

  • No detectable cross-reactivity with other relevant proteins

Interleukin-22 (IL-22), also known as ILTIF, is protein that in humans is encoded by the IL22 gene. IL-22 a member of a group of cytokines called the IL-10 family or IL-10 superfamily a class of potent mediators of cellular inflammatory responses. Using FISH, the IL22 gene is mapped to chromosome 12q15, close to the IFNG and the herpesvirus saimiri-induced AK155 genes. IL-22 can contribute to immune disease through the stimulation of inflammatory responses, S100s and defensins. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10. In some contexts, the pro-inflammatory versus tissue-protective functions of IL-22 are regulated by the often co-expressed cytokine IL-17A.