Order Entry
Belgium
ContactUsLinkComponent
Human IL-20 ELISA kit
Human IL-20 ELISA kit
Catalog # BIRBORB99227-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human IL-20 ELISA kit
Catalog # BIRBORB99227-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-20
  • Description:
    Human IL-20 ELISA Kit
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human IL-20 ELISA Kit for the quantitative detection of human IL-20 in cell culture supernates, serum and plasma (heparin, EDTA).

  • No detectable cross-reactivity with other relevant proteins

Interleukin-20 (IL-20) is a protein belonging to the IL-10 family of cytokines. IL-20 is produced by activated keratinocytes and monocytes and transmits an intracellular signal through two distinct cell-surface receptor complexes on keratinocytes and other epithelial cells. By radiation hybrid analysis, the IL20 gene was mapped to chromosome 1q32, where it is tightly linked to the IL10, IL19, and MDA7 genes within a 195-kb region, the IL10 family cytokine cluster. IL-20 regulates proliferation and differentiation of keratinocytes during inflammation, particularly inflammation associated with the skin. In addition, IL-20 also causes cell expansion of multipotential hematopoietic progenitor cells.