Order Entry
Belgium
ContactUsLinkComponent
 
Human furin ELISA kit
Catalog # BIRBORB96801-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human furin ELISA kit
Catalog # BIRBORB96801-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Human
  • Description:
    Human Furin ELISA Kit
  • Specificity:
    Natural and recombinant human Furin
  • Detection range:
    312 - 20000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human Furin ELISA Kit for the quantitative detection of human furin.

  • No detectable cross-reactivity with other relevant proteins

FurinFur, also known as PACE, is a protein that in humans is encoded by the FURIN gene. The protein encoded by this gene is an enzyme which belongs to the subtilisin-like proprotein convertase family. The FUR gene is located approximately 1 kb upstream of the FES gene on chromosome 15q25-q26. This gene is thought to play a role in tumor progression. Furin is one of the proteases responsible for the proteolytic cleavage of HIV envelope polyprotein precursor gp160 to gp120 and gp41 prior to viral assembly. Furin is enriched in the Golgi apparatus, where it functions to cleave other proteins into their mature or active forms. Expression of furin in T-cells is required for maintenance of peripheral immune tolerance.