Order Entry
Belgium
ContactUsLinkComponent
Human GDF-15 ELISA kit
Human GDF-15 ELISA kit
Catalog # BIRBORB96787-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human GDF-15 ELISA kit
Catalog # BIRBORB96787-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Human
  • Description:
    Human GDF-15 ELISA Kit
  • Specificity:
    Natural and recombinant human GDF-15
  • Detection range:
    31,2 - 2000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human GDF-15 ELISA Kit is a sandwich ELISA kit for the quantitative detection for human growth differentiation factor 15 (GDF-15).

  • No detectable cross-reactivity with other relevant proteins

GDF-15, also known as TGF-PL, MIC-1, PDF, PLAB, and PTGFB, is a protein belonging to the transforming growth factor beta superfamily that has a role in regulating inflammatory and apoptotic pathways in injured tissues and during disease processes. Using FISH, the MIC1 gene is mapped to 19p13.2-p13.1. Its expression in liver can be significantly up-regulated in during injury of organs such as liver, kidney, heart and lung. GDF15 showed increased expression and secretion during erythroblast maturation. GDF-15 functions as an anti-inflammatory cytokine by directly interfering with chemokine signaling and integrin activation.