Order Entry
Belgium
ContactUsLinkComponent
Mouse CTLA4 ELISA kit
Mouse CTLA4 ELISA kit
  BIRBORB96781-96
 :  Biorbyt
 :  
undefined
Mouse CTLA4 ELISA kit
  BIRBORB96781-96
 :  Biorbyt
 :  

 

  • Pk:
    96 Tests
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse CTLA4 ELISA Kit
  • Specificity:
    Natural and recombinant mouse CTLA4
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10 pg/ml

 

 

Mouse CTLA4 ELISA Kit is a sandwich ELISA kit for the quantitative detection for mouse CTLA4 (Cytotoxic T-Lymphocyte Antigen 4). CTLA4 is a protein receptor that down regulates the immune system.

  • No detectable cross-reactivity with other relevant proteins