ContactUsLinkComponent
Human HSP27 ELISA Kit
  BIRBORB78791-96
 :  Biorbyt
 :  
Human HSP27 ELISA Kit
  BIRBORB78791-96
 :  Biorbyt
 :  
  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human HSP27 ELISA Kit
  • Detection range:
    0,15 - 50 ng/ml
  • Sensitivity:
    150 pg/ml
Human HSP27 ELISA Kit ELISA kit is an in vitro ELISA for the quantitative measurement of human heat shock protein 27 (HSP27) in serum, plasma, cell culture supernatants and urine.
  • No detectable cross-reactivity with other relevant proteins
 : Kit includes HSP27 microplate (96 wells), coated with anti-human HSP27, wash buffer concentrate 20X (25 ml), standards (2 vials), assay diluent 5X (15 ml), detection antibody (2 vial), HRP-streptavidin concentrate 300X (200 µl), TMB one-step substrate reagent (12 ml), stop solution (8 ml).