Dipotassium hydrogen orthophosphate trihydrate, white crystalline powder, Reagent Grade
:16788-57-1
:7778-77-0
:K₂HPO₄·3H₂O
:MFCD00149926
:228.22 g/mol
:Ambient
- Identity Test:Passes
- FTIR:Conforms to Reference
- pH (5% aq soln):8,5 - 9,6
- Solubility (0,2 g/ml in water):Clear, colorless
Frequently Bought Together