L(+)-Potassium sodium tartrate tetrahydrate ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:6381-59-5
:
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989
:282.22 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together