Orders LinkContactUsLinkComponent
Hydrometers
Supplier:   GERING
Hydrometers
Supplier:  GERING
Certificates
Hydrometers
15 Options Available Below
Product Details & Documents
Set of 14 hydrometers calibrated at 20 °C.
 : Supplied in a velvet lined case.
Product Family Options
Product Information
  • Range
    Division
    Availability
    Price
  • 0,600 - 0,700 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,80 €
    35,80 €
    1 * 1 ST
       35,80 €
  • 0,700 - 0,800 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,80 €
    35,80 €
    1 * 1 ST
       35,80 €
  • 0,800 - 0,900 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,80 €
    35,80 €
    1 * 1 ST
       35,80 €
  • 0,900 - 1,000 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,80 €
    35,80 €
    1 * 1 ST
       35,80 €
  • 1,000 - 1,100 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,70 €
    36,70 €
    1 * 1 ST
       36,70 €
  • 1,100 - 1,200 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,70 €
    36,70 €
    1 * 1 ST
       36,70 €
  • 1,200 - 1,300 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,70 €
    36,70 €
    1 * 1 ST
       36,70 €
  • 1,300 - 1,400 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,10 €
    41,10 €
    1 * 1 ST
       41,10 €
  • 1,400 - 1,500 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,10 €
    41,10 €
    1 * 1 ST
       41,10 €
  • 1,500 - 1,600 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,10 €
    41,10 €
    1 * 1 ST
       41,10 €
  • 1,600 - 1,700 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,20 €
    46,20 €
    1 * 1 ST
       46,20 €
  • 1,700 - 1,800 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,20 €
    46,20 €
    1 * 1 ST
       46,20 €
  • 1,800 - 1,900 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,20 €
    46,20 €
    1 * 1 ST
       46,20 €
  • 1,900 - 2,000 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,20 €
    46,20 €
    1 * 1 ST
       46,20 €
  • 0,600 - 2,000 g/l
    0,001 g/ml
    Log in for availability
    Log in for availability
    1 * 14 ST
    1 * 14 ST