Sodium calibration standards for ISE measurement, Orion™
Frequently Bought Together
VWR®, Flash point reference standard, Pensky Martens
: VWR Chemicals
LAURYL ALCOHOL 1 * 1 g
: Merck
Imidacloprid 100 ng/µl 1 * 1 mL
: EHRENSTORFER
pH buffer solutions
: Hach
tert.-Butylhydroquinone certified 1 * 0,25 g
: EHRENSTORFER
VWR®, Total Base Number (TBN) standards
: VWR Chemicals