ContactUsLinkComponent
Cell culture media, powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Cell culture media, powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Catalog # ICNA091003122
Supplier:  MP Biomedicals
CAS Number:  
Cell culture media, powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Catalog # ICNA091003122
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Media Format:
    Powder
  • Pk:
    10 L
  • Description:
    BME (Modified), with Hanks' salts and L-glutamine, without sodium bicarbonate, 1×10 l
Specifications
About this item
Basal Medium Eagle is suited for the isolation and cultivation of non-fastidious and fastidious microorganisms from a variety of clinical and non-clinical specimens. It can also be used as a selective isolation medium by adding antimicrobial agents.