ContactUsLinkComponent
Detectable spatulas, SteriPlast®
Detectable spatulas, SteriPlast®
Catalog # 231-0574
Supplier:  BURKLE
undefined
Detectable spatulas, SteriPlast®
Catalog # 231-0574
Supplier:  BURKLE
Specifications
  • Colour:
    Blue
  • Length:
    263 mm
  • Pk:
    10
  • Blade length:
    150 mm
Specifications
About this item
These detectable SteriPlast® spatulas, made of blue PS, are produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. Ideal for sampling powders, granulates and pastes.
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
These spatulas are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.