ContactUsLinkComponent
Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Catalog # SIMELD3059
Supplier:  Industrial Physics
undefined
Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Catalog # SIMELD3059
Supplier:  Industrial Physics
Specifications
  • Description:
    Guide and reference photographs for steel surfaces prepared by wet abrasive blast cleaning
  • Pk:
    1
Specifications
About this item
This pictorial standard shows two uncoated, rusted steel surfaces (Conditions C and D) before and after wet abrasive blast cleaning. Photographs illustrate two degrees of cleaning (WAB 6 and WAB 10) for each initial condition with additional photos that depict the appearance of light, moderate, and heavy flash rust after cleaning. Also contains a written guide to the use of reference photographs and additional explanatory notes. Cleanliness requirements for conditions depicted are defined by the joint SSPC/NACE surface preparation specifications for commercial (SP 6/NACE 3) and near-white blast cleaning (SP 10/NACE 2). Photos of commercial and near white surfaces achieved by dry abrasive blast cleaning can be found in SSPC-VIS 1.