Orders LinkContactUsLinkComponent
Single gas detector
Single gas detector
Single gas detector, O₂
Catalog # SIMEDM6025
Supplier:   Industrial Physics
Single gas detector
Catalog # SIMEDM6025
Supplier:  Industrial Physics
Single gas detector
Single gas detector
Single gas detector, O₂
Price
272,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This gas clip detection equipment is designed for use in the coal mining industry.
  • Adjustable alarm set points
  • Real-time gas reading capability
  • Two-way infrared communication
Documents
Specifications
  • Parameter(s):
    O₂
  • Description:
    Single gas detector