ContactUsLinkComponent
Reference photographs for painted steel surfaces, SSPC-VIS 2
Reference photographs for painted steel surfaces, SSPC-VIS 2
Catalog # SIMELD3056
Supplier:  Industrial Physics
undefined
Reference photographs for painted steel surfaces, SSPC-VIS 2
Catalog # SIMELD3056
Supplier:  Industrial Physics
Specifications
  • Description:
    Method for evaluating degree of rusting on painted steel surfaces, SSPC-VIS 2
  • Pk:
    1
Specifications
About this item
SSPC-VIS 2 pictorial provides a method of evaluating degree of rusting on painted steel surfaces.
  • Series of full colour images
  • 27 black and white diagrams
  • Various degrees of spot, general and pinpoint rusting on painted steel surfaces
Text and tables provide a guide to the use of the reference photographs, scales and description of standard rust grades, comparison of SSPC, ASTM, ISO and other rust grade scales and other useful information.