ContactUsLinkComponent
Spatulas for foodstuffs, SteriPlast®
Spatulas for foodstuffs, SteriPlast®
Catalog # 231-0586
Supplier:  BURKLE
undefined
Spatulas for foodstuffs, SteriPlast®
Catalog # 231-0586
Supplier:  BURKLE
Specifications
  • Colour:
    Blue
  • Length:
    263 mm
  • Pk:
    10
  • Blade length:
    150 mm
Specifications
About this item
The SteriPlast® spatula for foodstuffs, made of PS, is produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The spatula is ideal for sampling powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.