ContactUsLinkComponent
Radius meter
Radius meter
Catalog # SIMEDI0201
Supplier:  Industrial Physics
undefined
Radius meter
Catalog # SIMEDI0201
Supplier:  Industrial Physics
Specifications
  • Range:
    1 - 7 mm
  • Description:
    Radius gauge of 20 sheets
  • Pk:
    1
Specifications
About this item
This radius meter provides round edges in outward exposed windows, doors and window frames, with a minimal radius.
Ordering information: Includes 20 sheets.