Calibration standards for thickness gauges
Supplier: Industrial Physics
Certificates
About this item
These coated metal calibration standards consist of a set of four calibration plates.
They are composed of an epoxy coating on steel or aluminium when many gauges are in use or when accuracy verification is performed frequently.
Certifications: ISO/QS-9000 and in-house quality control requirements.
Frequently Bought Together