Orders LinkContactUsLinkComponent
Sampling kits, SteriPlast
Sampling kits, SteriPlast
Supplier:   BURKLE
Sampling kits, SteriPlast
Supplier:  BURKLE
Certificates
Sampling kits, SteriPlast
Sampling kits, SteriPlast
2 Options Available Below
Product Details & Documents
The sampling set SteriPlast Kit consists of ten sample spatulas or ten sample scoops and ten SteriBag Premium tamper-proof sample bags. Thus the kit is the ideal combination for sampling where purity and sterility is essential.
  • All parts are sterilised by gamma rays
  • Suitable for use with foodstuffs
  • Scoops or spatulas are cleanroom manufactured and individually packed
  • Sample bag SteriBag Premium has an integrated sealing and is liquid-tight
  • Unique tracking code on both the bag and the tear-off field guarantees complete documentation and traceability
Delivery information: SteriPlast kit includes ten 50 ml scoops or ten spatulas (150 mm insertion depth) and ten 300 ml SteriBag Premium sample bags.
Product Family Options
Product Information
  • Capacity
    Colour
    Availability
    Price
  • 231-0591
    SteriPlast kit with 10 scoops and 10 sample bags
    SteriPlast kit with 10 scoops and 10 sample bags
    50 ml + 300 ml
    White
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 56,10 €
    56,10 €
    1 * 1 EA
       56,10 €
  • 231-0592
    SteriPlast kit with 10 spatulas (150 mm insertion depth) and 10 sample bags
    SteriPlast kit with 10 spatulas (150 mm insertion depth) and 10 sample bags
    300 ml
    White
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 52,50 €
    52,50 €
    1 * 1 EA
       52,50 €