Orders LinkContactUsLinkComponent
Surface profile comparators
Surface profile comparators
Surface profile comparators
Surface profile comparators
Surface comparators for determining roughness by touch and sight.
  • Five different grades of roughness in each comparator
  • Designed to be used with the illuminated magnifier
  • Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
 : ASTM D4417
Product Family Options
  • Description
  • Surface profile comparator for shot
    1 * 1 ST
    1 * 1 ST - 297,00 €
    297,00 €
    1 * 1 ST
       297,00 €
  • Surface profile comparator for grit
    1 * 1 ST
    1 * 1 ST - 297,00 €
    297,00 €
    1 * 1 ST
       297,00 €
  • Surface profile comparator for sand
    1 * 1 ST
    1 * 1 ST - 297,00 €
    297,00 €
    1 * 1 ST
       297,00 €
Accessories
  •  
    Description
  • SIMELD2054
    Surface profile disc holder and magnifier (5×)
    1 * 1 ST - 125,00 €