Order Entry
Belgium
ContactUsLinkComponent
 
Formaldehyde 24% in aqueous solution
Catalog # VWRK4094.9020
Supplier:  KLINIPATH
CAS Number:  
Formaldehyde 24% in aqueous solution
Catalog # VWRK4094.9020
Supplier:  KLINIPATH
CAS Number:  

Specifications

  • Pack type:
    Metal drum, tap system
  • Pk:
    20 L
  • Seq:
    5302

Specifications

About this item

 :
 :30.03 g/mol
UN:3334
ADR:9,
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274