Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Formaldehyde 4%

Irritant
Health
Supplier:   KLINIPATH
Formaldehyde 4%
Supplier:  KLINIPATH
Certificates

2 Options Available Below

Product Details & Documents

Formaldehyde 4% is approximately equal to Formalin 10%

  • pH 7
  • Phosphate buffered
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specifications for Product Family

Specification Test Results

  • Formaldehyde (HCHO) g/l:
    38 - 42
  • pH at 25 °C:
    6.8 - 7.2
  • Sodium (Na):
    61 - 71 mmol/l
Product Family Options

Product Information