

Xylene (mixture of isomers), pure
Supplier: KLINIPATH
Certificates
About this item
CAS Number:1330-20-7
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264
:106.17 g/mol
UN:1307
ADR:3,III
:GHS02
:GHS07
Related Information
Certificates
Search by Lot Numbersds_VWRK4055
SDS
spec_VWRK4055-9020 spec_VWRK4055-9020 spec_VWRK4055-9025 spec_VWRK4055-9025
Product specification
Frequently Bought Together