ContactUsLinkComponent
Temperature loggers, CurveX-3 basic oven logger
Temperature loggers, CurveX-3 basic oven logger
Supplier:  Industrial Physics
Certificates
undefined
Temperature loggers, CurveX-3 basic oven logger
Supplier:  Industrial Physics
Specifications
  • Catalog No:
  • SIMECX3010
  • SIMECX3005
  • Display:
  • Three multi-colours LED's
  • Three multi-colours LED's
  • Interface:
  • USB
  • USB
  • Battery life:
  • 1200 hour continuous use
  • 1200 hour continuous use
  • Recording intervals:
  • 1 s - 60 min
  • 1 s - 60 min
  • Weight:
  • 190 g
  • 190 g
  • W×D×H:
  • 100×85×16 mm
  • 100×85×16 mm
  • Data memory - No. of readings:
  • 10 batches with 16000 or 1 batch with 160000 readings
  • 10 batches with 16000 or 1 batch with 160000 readings
Specifications
Related Information
Probes
Product Information
Frequently Bought Together