Order Entry
Belgium
ContactUsLinkComponent
Fast RNA™ Pro Soil-Indirect Kit, MP Biomedicals
Fast RNA™ Pro Soil-Indirect Kit, MP Biomedicals
  ICNA116075050
 :  
undefined
Fast RNA™ Pro Soil-Indirect Kit, MP Biomedicals
  ICNA116075050
 :  

 

  • Pk:
    50 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Soil
  • Target molecule:
    RNA
  • Description:
    Fast RNA™ Pro Soil-Indirect Kit, 50×2 ml preps
  • Description:
    Fast RNA™ Pro Soil-Indirect Kit

 

 

FastRNA™ Pro Soil-Indirect Kit is used to isolate total RNA from organic material found in soil samples. This kit allows for the sample to be slurried with water or buffer in order to further separate biomass from the soil or environmental matrix, and the resulting supernatant is then filtered, lysed, and purified through the rest of the protocol. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.

The FastRNA™ Pro Soil-Indirect Kit is designed to efficiently isolate total RNA from organic material found in soil supernatants. Cellular material is washed away from soil particles, centrifuged, and homogenised by any FastPrep® instrument in impact-resistant 2 ml tubes of Lysing Matrix E. Total RNA is released into a protective solution, extracted, precipitated, and cleaned with the proprietary RNAMATRIX™ and a Quick-Clean Spin Filter. Total RNA prepared with this kit is suitable for RT-PCR analysis and other downstream applications.

For research purposes only.

 : Kit contains RNApro™ Solution; Lysing Matrix E; Inhibitor Removal Solution; DEPC-treated H₂O; RNAMATRIX® Binding Solution; RNAMATRIX® Slurry; RNAMATRIX® Wash Solution; and Quick-Clean Spin Filters.