ContactUsLinkComponent
Deuterium lamps
Deuterium lamps
Catalog # VARI393570502
Supplier:  VARIAN
Deuterium lamps
Catalog # VARI393570502
Supplier:  VARIAN
Specifications
  • For:
    Varian - Prostar 330 DAD
  • Description:
    Deuterium lamp, D2
  • Pk:
    1
Specifications
About this item
Deuterium lamp for Hitachi and Varian detectors. Lifetime 1000 hrs.
Suitable for:
Merck / Hitachi - L-2455 DAD, L-2500, L-3000, L-4000, L-4200, L-4225, L-4250, L-4500, L-4720, L-6000, L-7400, L-7420, L-7450, L-7450A, L-7455, LaChrom, L2400LCE, L2420LCE, L2450
Varian - ProStar 330 DAD